missing translation for 'onlineSavingsMsg'
Learn More

Fumaryl Chloride 95.0+%, TCI America™

Catalog No. F0152500G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F0152500G 500 g
F015225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. F0152500G Supplier TCI America Supplier No. F0152500G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 627-63-4
Molecular Formula C4H2Cl2O2
Molecular Weight (g/mol) 152.958
MDL Number MFCD00000733
InChI Key ZLYYJUJDFKGVKB-OWOJBTEDSA-N
Synonym fumaryl chloride, fumaroyl dichloride, fumaroyl chloride, fumarylchlorid, chlorure de fumaryle, fumarylchlorid czech, 2-butenedioyl dichloride, e, 2-butenedioyl dichloride, 2e, dichlorid kyseliny fumarove, fumaryl dichloride
PubChem CID 5325504
IUPAC Name (E)-but-2-enedioyl dichloride
SMILES C(=CC(=O)Cl)C(=O)Cl

Specifications

Melting Point -2°C
Color Yellow
Boiling Point 164°C
Quantity 500 g
UN Number 1780
Formula Weight 152.96
Percent Purity ≥95.0% (GC,T)
Physical Form Liquid
Chemical Name or Material Fumaryl Chloride
EINECSNumber (2)-1096
RTECSNumber LT2800000
TSCA Yes