Learn More
Fumaryl chloride, 95%
CAS: 627-63-4 | C4H2Cl2O2 | 152.96 g/mol
$316.07 - $316.07
Chemical Identifiers
| CAS | 627-63-4 |
|---|---|
| Molecular Formula | C4H2Cl2O2 |
| Molecular Weight (g/mol) | 152.96 |
| MDL Number | MFCD00000733 |
| InChI Key | ZLYYJUJDFKGVKB-OWOJBTEDSA-N |
| Synonym | fumaryl chloride, fumaroyl dichloride, fumaroyl chloride, fumarylchlorid, chlorure de fumaryle, fumarylchlorid czech, 2-butenedioyl dichloride, e, 2-butenedioyl dichloride, 2e, dichlorid kyseliny fumarove, fumaryl dichloride |
| PubChem CID | 5325504 |
| IUPAC Name | (E)-but-2-enedioyl dichloride |
| SMILES | C(=CC(=O)Cl)C(=O)Cl |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
AC165971000
|
Thermo Scientific Chemicals
165971000 |
100 mL | Glass bottle |
N/A
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 627-63-4 | |
| 152.96 | |
| ZLYYJUJDFKGVKB-OWOJBTEDSA-N | |
| 5325504 | |
| C(=CC(=O)Cl)C(=O)Cl |
| C4H2Cl2O2 | |
| MFCD00000733 | |
| fumaryl chloride, fumaroyl dichloride, fumaroyl chloride, fumarylchlorid, chlorure de fumaryle, fumarylchlorid czech, 2-butenedioyl dichloride, e, 2-butenedioyl dichloride, 2e, dichlorid kyseliny fumarove, fumaryl dichloride | |
| (E)-but-2-enedioyl dichloride |
Specifications
| 627-63-4 | |
| Yellow | |
| 161.0°C to 164.0°C | |
| Authentic | |
| Glass bottle | |
| 1.4980 to 1.5010 | |
| MFCD00000733 | |
| 02, 743 | |
| fumaryl chloride, fumaroyl dichloride, fumaroyl chloride, fumarylchlorid, chlorure de fumaryle, fumarylchlorid czech, 2-butenedioyl dichloride, e, 2-butenedioyl dichloride, 2e, dichlorid kyseliny fumarove, fumaryl dichloride | |
| ZLYYJUJDFKGVKB-OWOJBTEDSA-N | |
| (E)-but-2-enedioyl dichloride | |
| 5325504 | |
| 95% | |
| Fumaryl chloride, 95% |
| -2.0°C | |
| 1.4100g/mL | |
| 73°C | |
| 94% min. (GC) | |
| C4H2Cl2O2 | |
| ClCOCH=CHCOCl | |
| 100 mL | |
| 1.41 | |
| Solubility in water: decomposes | |
| C(=CC(=O)Cl)C(=O)Cl | |
| 152.96 | |
| 152.96 | |
| Liquid |
Safety and Handling
GHS H Statement:
Harmful if swallowed.
Causes serious eye damage.
Harmful if inhaled.
Harmful in contact with skin.
Causes severe skin burns and eye damage.
Contact with water liberates toxic gas.
GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.
Store in a dry place. Store in a closed container.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Danger
EINECSNumber : 211-005-4
RTECSNumber : LT2800000
TSCA : TSCA
RUO – Research Use Only