missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fumaronitrile 98.0+%, TCI America™
Supplier: TCI America F007225G
Spécifications
| Fumaronitrile | |
| 97°C | |
| 186°C | |
| MFCD00001928 | |
| 3439 | |
| KYPOHTVBFVELTG-OWOJBTEDSA-N | |
| (E)-but-2-enedinitrile | |
| 637930 | |
| ≥98.0% (GC) |
| 764-42-1 | |
| White-Yellow | |
| C4H2N2 | |
| 25 g | |
| fumaronitrile, fumarodinitrile, furmaronitrile, fumaric nitrile, 2-butenedinitrile, e, 2-butenedinitrile, 2e, trans-1,2-dicyanoethylene, trans-1,2-dicyanoethene, 2e-but-2-enedinitrile, e-1,2-dicyanoethylene | |
| C(=CC#N)C#N | |
| 78.074 | |
| 78.07 | |
| Crystalline Powder |
Identifiants chimiques
| 764-42-1 | |
| 78.074 | |
| KYPOHTVBFVELTG-OWOJBTEDSA-N | |
| 637930 | |
| C(=CC#N)C#N |
| C4H2N2 | |
| MFCD00001928 | |
| fumaronitrile, fumarodinitrile, furmaronitrile, fumaric nitrile, 2-butenedinitrile, e, 2-butenedinitrile, 2e, trans-1,2-dicyanoethylene, trans-1,2-dicyanoethene, 2e-but-2-enedinitrile, e-1,2-dicyanoethylene | |
| (E)-but-2-enedinitrile |
Sécurité et manipulation
missing translation for 'einecsNumber' : (9)-1274
missing translation for 'rtecsNumber' : LT2300000
missing translation for 'tsca' : Yes