missing translation for 'onlineSavingsMsg'
Learn More

Fumaronitrile, 98%, Thermo Scientific™

Chemical Identifiers

CAS 764-42-1
Molecular Formula C4H2N2
Molecular Weight (g/mol) 78.06
MDL Number MFCD00001928
InChI Key KYPOHTVBFVELTG-OWOJBTEDSA-N
Synonym fumaronitrile, fumarodinitrile, furmaronitrile, fumaric nitrile, 2-butenedinitrile, e, 2-butenedinitrile, 2e, trans-1,2-dicyanoethylene, trans-1,2-dicyanoethene, 2e-but-2-enedinitrile, e-1,2-dicyanoethylene
PubChem CID 637930
IUPAC Name (E)-but-2-enedinitrile
SMILES C(=CC#N)C#N
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC157400050
SDS View Documents
Thermo Scientific Chemicals
157400050
5g Glass bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC157400250
SDS View Documents
Thermo Scientific Chemicals
157400250
25g Glass bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC157401000
SDS View Documents
Thermo Scientific Chemicals
157401000
100g Glass bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

764-42-1
78.06
KYPOHTVBFVELTG-OWOJBTEDSA-N
637930
C(=CC#N)C#N
C4H2N2
MFCD00001928
fumaronitrile, fumarodinitrile, furmaronitrile, fumaric nitrile, 2-butenedinitrile, e, 2-butenedinitrile, 2e, trans-1,2-dicyanoethylene, trans-1,2-dicyanoethene, 2e-but-2-enedinitrile, e-1,2-dicyanoethylene
(E)-but-2-enedinitrile

Specifications

764-42-1
Authentic
Glass bottle
NCCH=CHCN
5g
fumaronitrile, fumarodinitrile, furmaronitrile, fumaric nitrile, 2-butenedinitrile, e, 2-butenedinitrile, 2e, trans-1,2-dicyanoethylene, trans-1,2-dicyanoethene, 2e-but-2-enedinitrile, e-1,2-dicyanoethylene
C(=CC#N)C#N
78.06
78.06
Fumaronitrile
186.0°C
97.5% min. (GC)
C4H2N2
MFCD00001928
02, I, 302
KYPOHTVBFVELTG-OWOJBTEDSA-N
(E)-but-2-enedinitrile
637930
98%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Toxic in contact with skin.
Toxic if swallowed.

GHS P Statement
Do not eat,drink or smoke when using this product.
Wash face,hands and any exposed skin thoroughly after handling.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Avoid breathing dust/fume/gas/mis

GHS Signal Word: Danger

EINECSNumber : 212-122-3

SDS
Documents

Documents

RUO – Research Use Only