missing translation for 'onlineSavingsMsg'
Learn More

Fumaronitrile, 95%

Catalog No. AC157401000
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC157401000 100 g Glass bottle
AC157400050 5 g Glass bottle
AC157400250 25 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC157401000 Supplier Thermo Scientific Chemicals Supplier No. 157401000
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 764-42-1 | C4H2N2 | 78.06 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

Chemical Identifiers

CAS 764-42-1
Molecular Formula C4H2N2
Molecular Weight (g/mol) 78.06
MDL Number MFCD00001928
InChI Key KYPOHTVBFVELTG-OWOJBTEDSA-N
Synonym fumaronitrile, fumarodinitrile, furmaronitrile, fumaric nitrile, 2-butenedinitrile, e, 2-butenedinitrile, 2e, trans-1,2-dicyanoethylene, trans-1,2-dicyanoethene, 2e-but-2-enedinitrile, e-1,2-dicyanoethylene
PubChem CID 637930
IUPAC Name (E)-but-2-enedinitrile
SMILES C(=CC#N)C#N

Specifications

Melting Point 95.0°C to 98.0°C
Color White
Boiling Point 186.0°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula NCCH=CHCN
Quantity 100 g
Beilstein 02, I, 302
Formula Weight 78.06
Percent Purity 98%
Physical Form Crystals
Chemical Name or Material Fumaronitrile
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Toxic in contact with skin.
Toxic if swallowed.
Precautionary Statement GHS P Statement
Do not eat,drink or smoke when using this product.
Wash face,hands and any exposed skin thoroughly after handling.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Avoid breathing dust/fume/gas/mis
EINECSNumber 212-122-3

RUO – Research Use Only