missing translation for 'onlineSavingsMsg'
Learn More

Fumaronitrile, 98+%

Catalog No. p-7046548
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1491106 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAL1491106 Supplier Thermo Scientific Chemicals Supplier No. L1491106
Only null left

CAS: 764-42-1 | C4H2N2 | 78.074 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 764-42-1
Molecular Formula C4H2N2
Molecular Weight (g/mol) 78.074
MDL Number MFCD00001928
InChI Key KYPOHTVBFVELTG-OWOJBTEDSA-N
Synonym fumaronitrile, fumarodinitrile, furmaronitrile, fumaric nitrile, 2-butenedinitrile, e, 2-butenedinitrile, 2e, trans-1,2-dicyanoethylene, trans-1,2-dicyanoethene, 2e-but-2-enedinitrile, e-1,2-dicyanoethylene
PubChem CID 637930
IUPAC Name (E)-but-2-enedinitrile
SMILES C(=CC#N)C#N

Specifications

Melting Point 94°C to 99°C
Density 0.94
Boiling Point 186°C
Odor Recognizable
Quantity 5 g
UN Number UN3439
Beilstein 969245
Formula Weight 78.07
Percent Purity ≥98%
Chemical Name or Material Fumaronitrile
Hazard Category H301+H331
Hazard Statement GHS H Statement
H301
Toxic if swallowed.
Precautionary Statement P261-P264b-P270-P271-P301+P310-P304+P340-P311-P330-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITRILES, SOLID, TOXIC, N.O.S.
EINECSNumber 212-122-3
RTECSNumber LT2300000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only