missing translation for 'onlineSavingsMsg'
Learn More

Fumaraldehyde Bis(dimethyl Acetal) 90.0+%, TCI America™

Catalog No. F03295G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F03295G 5 g
F03291G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. F03295G Supplier TCI America Supplier No. F03295G
Only null left

Chemical Identifiers

CAS 6068-62-8
Molecular Formula C8H16O4
Molecular Weight (g/mol) 176.212
MDL Number MFCD00075507
InChI Key ZFGVCDSFRAMNMT-AATRIKPKSA-N
Synonym fumaraldehyde bis dimethyl acetal, e-1,1,4,4-tetramethoxybut-2-ene, fumaraldehyde bis dimethylacetal, 2-butenedial bis dimethyl acetal, 1,1,4,4-tetramethoxy-trans-2-butene, trans-1,1,4,4-tetramethoxy-2-butene, 1,1,4,4-tetramethoxy-but-2-ene
PubChem CID 5862719
IUPAC Name (E)-1,1,4,4-tetramethoxybut-2-ene
SMILES COC(C=CC(OC)OC)OC

Specifications

Color Colorless
Boiling Point 103°C
Quantity 5 g
Formula Weight 176.21
Percent Purity ≥90.0% (GC)
Physical Form Liquid
Chemical Name or Material Fumaraldehyde Bis(dimethyl Acetal)