missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fumaraldehyde Bis(dimethyl Acetal) 90.0+%, TCI America™
Specifications
Specifications
| Color | Colorless |
| Boiling Point | 103°C |
| Quantity | 1 g |
| Formula Weight | 176.21 |
| Percent Purity | ≥90.0% (GC) |
| Physical Form | Liquid |
| Chemical Name or Material | Fumaraldehyde Bis(dimethyl Acetal) |
Safety and Handling
| TSCA | No |