missing translation for 'onlineSavingsMsg'
Learn More

Fumagillin, 95%, Thermo Scientific Chemicals

Catalog No. AAJ67045MCR
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ67045MCR 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAJ67045MCR Supplier Thermo Scientific Chemicals Supplier No. J67045MCR

 

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 23110-15-8
Molecular Formula C26H34O7
Molecular Weight (g/mol) 458.551
MDL Number MFCD03990453
InChI Key NGGMYCMLYOUNGM-HVXGILEMSA-N
Synonym fumagillin, fugillin, fumidil, fumagillin dch, amebacilin, fumadil b, --fumagillin, 2e,4e,6e,8e-10-3r,4s,5r,6r-5-methoxy-4-2r,3r-2-methyl-3-3-methylbut-2-en-1-yl oxiran-2-yl-1-oxaspiro 2.5 octan-6-yl oxy-10-oxodeca-2,4,6,8-tetraenoic acid, 2e,4e,6e,8e-2,4,6,8-decatetraenedioic acid 1-3r,4s,5s,6r-5-methoxy-4-2r,3r-2-methyl-3-3-methyl-2-buten-1-yl-2-oxiranyl-1-oxaspiro 2.5 oct-6-yl ester, 2,4,6,8-decatetraenedioic acid mono 4-1,2-epoxy-1,5-dimethyl-4-hexenyl-5-methoxy-1-oxaspiro 2.5 oct-6-yl ester
PubChem CID 71317005
IUPAC Name (2E,4E,6E,8E)-10-[[(3R,4S,5R,6R)-5-methoxy-4-[(2R,3R)-2-methyl-3-(3-methylbut-2-enyl)oxiran-2-yl]-1-oxaspiro[2.5]octan-6-yl]oxy]-10-oxodeca-2,4,6,8-tetraenoic acid
SMILES CC(=CCC1C(O1)(C)C2C(C(CCC23CO3)OC(=O)C=CC=CC=CC=CC(=O)O)OC)C

Specifications

Melting Point 194°C to 195°C
Color White to Yellow
Odor Odorless
Quantity 1 mg
Sensitivity Air sensitive
Merck Index 14,4286
Formula Weight 458.55
Percent Purity 95%
Physical Form Solid
Chemical Name or Material Fumagillin
Hazard Category H302
Hazard Statement GHS H Statement
H302
Harmful if swallowed.
Precautionary Statement P264b-P270-P301+P312-P330-P501c
EINECSNumber 245-433-8
RTECSNumber HE1750000
TSCA No
storageNote1 -30° to -10°C
Recommended Storage Store at -20°C; Store under Argon

RUO – Research Use Only