missing translation for 'onlineSavingsMsg'
Learn More

Honeywell™ North™ RU6500 Series Full-Facepiece Air-Purifying Respirators

Catalog No. 19085553
Change view
Click to view available options
Size:
Small
Medium
Large
Includes:
Head Strap
Head Strap and Mesh Head Net
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size Includes
19085553 Medium Head Strap
19085554 Large Head Strap
19085555 Small Head Strap and Mesh Head Net
19085557 Large Head Strap and Mesh Head Net
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 19085553 Supplier Honeywell™ Supplier No. RU65001M

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Requires

Compatible with North™ N-Series cartridges, filters and accessories.

Features a polycarbonate lens with a wide field of vision for maximized peripheral vision plus a downward view.

  • Silicone seal provides secure facepiece seal, creating a barrier against contaminants
  • Silicone nose cup is soft and comfortable for all day wear
  • Wide viewing area provides distortion free visibility and optical clarity
  • Polycarbonate lens meet ANSI Z.87 for high impact
  • Facepiece with 5-point strap includes center strap to help position and hold the mask for a more secure fit; offers quick, easy adjustments
  • Facepiece with optional industrial nylon mesh head net (19-085-555, 556, 557) provides another choice for user comfort preference; can be used under hard hat
  • Threaded cartridge connector secures equipment to avoid accidental disconnections
  • Can be adapted to side- or front-mounted PAPR or CF-SAR; compatible with PA700 Series PAPR; PR500 Series Mask-Mounted PAPR; CF2000 Series SAR; and CF4000 Series Mask-Mounted SAR

Specifications

ANSI Standard ANSI Z87.1
Color Black
Material Silicone
Product Type Full Facepiece Respirator
Size Medium
Facepiece Material Silicone
For Use With (Equipment) North™ N-Series line of cartridges, filters, and cartridge/filter combinations
Includes Head Strap
Material (Lens) Polycarbonate
Model RU65001M
Suspension Type 5-point
Show More Show Less