missing translation for 'onlineSavingsMsg'
Learn More

CleanSpace™ Full Face Mask Orinasal Inner Mask Set for PAPR Assembly

Catalog No. 17800643
Change view
Click to view available options
Size:
Medium/Large
Small
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
17800643 Medium/Large
17800642 Small
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 17800643 Supplier CleanSpace™ Supplier No. PAF1027
Only null left

CleanSpace™ Full Face Mask Orinasals designed to be used with the CleanSpace™ Full Face Mask are made of silicone and polycarbonate.

  • Used with the revolutionary CleanSpace™ full face mask
  • Designed for comfort over long periods
  • Allows breathability and prevents fogging
  • Easy to wash and quick drying
  • Designed for long wear in harsh environments
  • Easy and quick replacement
  • Cleaning: Machine washable or use in warm soapy water. Do not use solvents (turpentine or acetone), hot water, bleaching or chemical agents

Specifications

Certifications/Compliance AS/NZS1716: 2012, AS/ NZS 1337.1:2010 High Impact Resistance
Certifications/Compliance (Europe) EN 12942
Certifications (EN) EN 12942
Product Type PAPR Full Face Mask
Size Medium/Large
Facepiece/Headtop Model PAPR full face mask only
For Use With (Application) Welding, woodworking, manufacturing, smelting, construction, recycling plants, emergency services, mining, agriculture, processing plants, grinding
For Use With (Equipment) PAF-1014, PAF-1106
Model PAF-1027
Storage Requirements Store at -10°C to 55°C
Dimensions (L x W x H) 140 x 110 x 90 mm
Weight (Metric) 6.5 g
Show More Show Less