missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fuchsin-Carbol, Ziehl-Neelsen Formulation, for Staining Acid Fast Organisms, Ricca Chemical
Staining Acid Fast Organisms
$140.25 - $279.91
Specifications
| Chemical Name or Material | Fuchsin-Carbol |
|---|---|
| CAS | 7732-18-5,64-17-5,108-95-2,67-56-1,569-61-9 |
| CAS Min % | 87.0,6.95,3.91,0.26 |
| CAS Max % | 90.0,7.23,4.07,0.27 |
| Grade | Laboratory |
Description
- Ziehl-Neelsen. Requires special barrier pack at additional charge for shipment via UPS Air
- Glass/Poisonous Liquid
Specifications
| Fuchsin-Carbol | |
| 87.0,6.95,3.91,0.26 | |
| Laboratory | |
| Dark Red |
| 7732-18-5,64-17-5,108-95-2,67-56-1,569-61-9 | |
| 90.0,7.23,4.07,0.27 | |
| Amber Glass Bottle |