missing translation for 'onlineSavingsMsg'
Learn More

Cupric Nitrate, Hydrate, Crystal, ACS, 98-102%, Spectrum™ Chemical

Catalog No. 18610011
Change view
Click to view available options
Quantity:
5 g
25 g
125 g
500 g
2.5 kg
12 kg
45 kg
Packaging:
Amber Glass Bottle
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18610011 45 kg Amber Glass Bottle
18610005 5 g Amber Glass Bottle
18610006 25 g Amber Glass Bottle
18610007 125 g Amber Glass Bottle
18610008 500 g Amber Glass Bottle
18610009 2.5 kg Amber Glass Bottle
18610010 12 kg Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Catalog No. 18610011 Supplier Spectrum Chemical Mfg Cor Supplier No. C140545KGBL
Get Up to $500 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 19004-19-4 | 232.59 g/mol

Chemical Identifiers

CAS 19004-19-4

Specifications

CAS Max % 1
Physical Form Crystalline Powder or Needles
Quantity 45 kg
Molecular Weight (g/mol) 232.59
Percent Purity 98 to 102%
Grade ACS/Reagent
Packaging Amber Glass Bottle
Chemical Name or Material Cupric Nitrate, Hydrate, Crystal

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.