missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fructose, Granular, FCC, 98-102%, Spectrum™ Chemical
F1092, 57-48-7, C6H12O6
$980.37 - $5,672.77
Chemical Identifiers
| CAS | 57-48-7 |
|---|---|
| Molecular Formula | C6H12O6 |
| Molecular Weight (g/mol) | 180.16 |
| InChI Key | BJHIKXHVCXFQLS-UYFOZJQFSA-N |
| IUPAC Name | (3S,4R,5R)-1,3,4,5,6-pentahydroxyhexan-2-one |
| SMILES | OC[C@@H](O)[C@@H](O)[C@H](O)C(=O)CO |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
18602856
|
Spectrum Chemical Mfg Cor
F109225KG |
2.5 kg | Poly Pail |
N/A
|
|
||||
|
18602855
|
Spectrum Chemical Mfg Cor
F1092500GMCS6 |
6 x 500 g | Poly Bottle |
N/A
|
|
||||
|
18602857
|
Spectrum Chemical Mfg Cor
F109225KGCS4 |
4 x 2.5 kg | Poly Pail |
N/A
|
|
||||
|
18602858
|
Spectrum Chemical Mfg Cor
F109212KG |
12 kg | Poly Pail |
N/A
|
|
||||
|
18602859
|
Spectrum Chemical Mfg Cor
F109245KGBL |
45 kg | Fiber Drum |
N/A
|
|
||||
Description
Spectrum™ Chemical Fructose, Granular, FCC is a white, odorless crystalline solid that is extremely sweet and is the most water soluble of all sugars. It is used commercially as a sweetener in food and beverages due to its high sweetness. The FCC grade meets the requirements of the Food Chemical Codex indicates and is suitable for all food, beverage and nutritional supplement applications. Spectrum Chemical offers over 300 Food grade chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.Chemical Identifiers
| 57-48-7 | |
| 180.16 | |
| (3S,4R,5R)-1,3,4,5,6-pentahydroxyhexan-2-one |
| C6H12O6 | |
| BJHIKXHVCXFQLS-UYFOZJQFSA-N | |
| OC[C@@H](O)[C@@H](O)[C@H](O)C(=O)CO |
Specifications
| 57-48-7 | |
| 0.5% | |
| Poly Pail | |
| 2.5 kg | |
| OC[C@@H](O)[C@@H](O)[C@H](O)C(=O)CO | |
| 180.16 | |
| FCC |
| 100% | |
| 0.5% | |
| C6H12O6 | |
| BJHIKXHVCXFQLS-UYFOZJQFSA-N | |
| (3S,4R,5R)-1,3,4,5,6-pentahydroxyhexan-2-one | |
| 98 to 102% |