missing translation for 'onlineSavingsMsg'
Learn More

Fructose Standard, 600 ppm C6H12O6, Ricca Chemical

Catalog No. R3218060100
Change view
Click to view available options
Packaging:
Natural Poly Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Packaging
R3218060100 Natural Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. R3218060100 Supplier Ricca Chemical Company Supplier No. R3218060100A

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 57-48-7
Molecular Formula C6H12O6
Molecular Weight (g/mol) 180.156
InChI Key BJHIKXHVCXFQLS-UYFOZJQFSA-N
PubChem CID 5984
ChEBI CHEBI:48095
IUPAC Name (3S,4R,5R)-1,3,4,5,6-pentahydroxyhexan-2-one
SMILES C(C(C(C(C(=O)CO)O)O)O)O

Specifications

Packaging Natural Poly Bottle
Chemical Name or Material Fructose Standard, 600ppm