missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fructose Standard, 20,000 ppm C6H12O6, Ricca Chemical
Supplier: Ricca Chemical Company R3219020100A
Specifications
| Fructose Standard, 20000ppm | |
| 57-48-7 | |
| BJHIKXHVCXFQLS-UYFOZJQFSA-N | |
| (3S,4R,5R)-1,3,4,5,6-pentahydroxyhexan-2-one | |
| 5984 |
| Natural Poly Bottle | |
| C6H12O6 | |
| C(C(C(C(C(=O)CO)O)O)O)O | |
| 180.156 | |
| CHEBI:48095 |
Chemical Identifiers
| 57-48-7 | |
| 180.156 | |
| 5984 | |
| (3S,4R,5R)-1,3,4,5,6-pentahydroxyhexan-2-one |
| C6H12O6 | |
| BJHIKXHVCXFQLS-UYFOZJQFSA-N | |
| CHEBI:48095 | |
| C(C(C(C(C(=O)CO)O)O)O)O |