missing translation for 'onlineSavingsMsg'
Learn More

Fructose Standard, 1500 ppm C6H12O6, Ricca Chemical

Supplier:  Ricca Chemical Company R3218150100A

Catalog No. R3218150100

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Explore available promotions

Description

Description

Specifications

Specifications

Fructose Standard, 1500ppm
57-48-7
BJHIKXHVCXFQLS-UYFOZJQFSA-N
(3S,4R,5R)-1,3,4,5,6-pentahydroxyhexan-2-one
5984
Natural Poly Bottle
C6H12O6
C(C(C(C(C(=O)CO)O)O)O)O
180.156
CHEBI:48095

Chemical Identifiers

57-48-7
180.156
5984
(3S,4R,5R)-1,3,4,5,6-pentahydroxyhexan-2-one
C6H12O6
BJHIKXHVCXFQLS-UYFOZJQFSA-N
CHEBI:48095
C(C(C(C(C(=O)CO)O)O)O)O
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions