missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Fructosamine-3-kinase-related Recombinant Protein Antigen
Description
A recombinant protein antigen with a N-terminal His6-ABP tag corresponding to human FN3KRP. The Fructosamine-3-kinase-related Recombinant Protein Antigen has been validated for the following applications: Antibody Competition.
Specifications
Specifications
| Gene ID (Entrez) | 79672 |
| Product Line | Novus |
| Purity | >80% by SDS-PAGE and Coomassie blue staining |
| Common Name | EC 2.7.1.-, FLJ12171, FN3KL, FN3K-related protein, FN3K-RP, fructosamine 3 kinase related protein, Fructosamine-3-kinase-related protein, ketosamine-3-kinase |
| Content And Storage | Store at -20°C. Avoid freeze-thaw cycles. |
| Gene Symbol | FN3KRP |
| Molecular Weight (g/mol) | 28 kDa |
| Quantity | 100 μL |
| Source | E. coli |
| Specific Reactivity | This recombinant antigen is only intended to be used as a blocking agent to confirm antibody specificity with the corresponding antibody, catalog number NBP2-54164. It is purified by IMAC chromatography, and the expected concentration is greater than 0.5 mg/ml |