Learn More
Mopec Front Filler With Toe Kick, Sitting Height,Adjustable
Description
Construction
- (1) Sitting height base filler with toekick
- Toe kick portion at the bottom is included
Materials
- 304 stainless steel construction with a polished No. 4 finish
Dimensions
- LCFFL-ADJS: 18 in. L x 1 in. W x 27.1 in. H
Specifications
Specifications
| Product Type | Front Filler |