missing translation for 'onlineSavingsMsg'
Learn More

Sodium Oxalate, Powder, Primary Standard, Reagent Special, ACS, 99.95-105%, Spectrum™ Chemical

Catalog No. 18606683
Change view
Click to view available options
Quantity:
125 g
Packaging:
Poly Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18606683 125 g Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 18606683 Supplier Spectrum Chemical Mfg Cor Supplier No. S1369125GM
Get Up to $300 in Additional Chemicals
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

S1369, 62-76-0, C2Na2O4

Spectrum™ Chemical Sodium Oxalate, Powder, Primary Standard, Reagent Special, ACS , also known as disodium oxalate, is used as a primary standard for standardizing potassium permanganate solutions. As an ACS grade quality reagent, its chemical specifications are the de facto standards for chemicals used in many high-purity applications and typically designate the highest quality chemical available for laboratory use. Spectrum Chemical manufactured Reagent ACS grade products meet the toughest regulatory standards for quality and purity.

Chemical Identifiers

CAS 62-76-0
Molecular Formula C2Na2O4
Molecular Weight (g/mol) 134.00
MDL Number MFCD00012465
InChI Key ZNCPFRVNHGOPAG-UHFFFAOYSA-L
IUPAC Name disodium oxalate
SMILES [Na+].[Na+].[O-]C(=O)C([O-])=O

Specifications

CAS Max % 1
Physical Form Powder, Needles or Crystals
Quantity 125 g
Percent Purity 99.95 to 100.05%
Grade ACS/Reagent
Loss on Drying 0.0001
Packaging Poly Bottle
Chemical Name or Material Sodium Oxalate, Powder, Primary Standard