Learn More
Chemglass Inc Glass Frit, 40-60 Micron
Supplier: Chemglass Inc CLS1437G1
Made of glass and easily attaches to one ofdip tube assemblies with silicone tubing (not provided) to help increase aeration.Porosity is 40-60 micron.Can be autoclaved.Frit assembly 50mm.Stem OD 8mm.