Learn More
United Scientific™ Friction Rods and Pads
Ideal tools for demonstrating static electricity.
Supplier: United Scientific™ FRHG12
Description
- Friction rods and pads made from various materials.
- All the essential materials for student exploration in electrostatics
- Topics include static electricity, polarity, and friction studies
- Use a variety of friction rods, friction pads, and pith balls
- The combination of materials will determine the charge distribution after contact is made
Chemical Identifiers
| Friction Rods and Pads | |
| Friction Demonstrations |
Specifications
| Friction Rods and Pads | |
| Glass | |
| 12 in. |
| Hollow Rod | |
| Friction Demonstrations |