missing translation for 'onlineSavingsMsg'
Learn More

United Scientific™ Friction Cubes Pack of 5

Catalog No. S000868
Change view
Click to view available options
Grade:
7 to 12
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Grade
S000868 7 to 12
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Prop 65
Catalog No. S000868 Supplier United Scientific™ Supplier No. FRCUBE5

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Prop 65

Friction Cubes Set of 5 explores how the different surfaces affect the frictional force on a moving object.

Explore how the different surfaces affect the frictional force on a moving object. This square wooden cube has four different faces: bare wood, paper, vinyl and sandpaper. You can explore the value for static and kinetic friction by either pulling on the block or allowing it to slide slowly down an incline plane. Required but not included are masses, incline plane and a spring scale.

  • Four different surfaces
  • Large 2 in. cube
  • Meets NGSS Standards
  • Includes activity guide

Specifications

Grade 7 to 12
Class Size 1 Lab Group
Product Type Friction Cubes