missing translation for 'onlineSavingsMsg'
Learn More

United Scientific™ Friction Board with Blocks

Catalog No. S000854
Change view
Click to view available options
Grade:
5+
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Grade
S000854 5+
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Prop 65
Catalog No. S000854 Supplier United Scientific™ Supplier No. FRCBRD

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Prop 65

Our Friction Board with Blocks demonstrates the difference in force required to drag blocks across different surfaces.

The force of friction acts upon any moving object. The surfaces that are in contact with each other directly affect the frictional force that the object experiences. When the blocks are dragged across different surfaces, the force needed to keep the block moving varies from one surface to another. Adding masses on top of the blocks adds an additional variable for investigation. The four surfaces are: cardboard, sandpaper (220 grit), cork, and vinyl. Required but not included are a spring scale and additional masses.

  • Each section measures 8 in. long x 3 in. wide.
  • Blocks measure 1.75 x 1.75 x 0.75 in.
  • Introduce how surface texture affects friction.
  • Four different surfaces.
  • Includes activity guide

Specifications

Grade 5+
Class Size 1 Lab Group
Product Type Friction Board