missing translation for 'onlineSavingsMsg'
Learn More

Vileda Professional™ Roll-O-Matic™ Sponge Mop Refills

Catalog No. 190321020
Change view
Click to view available options
Material:
CE Universal Stainless Steel
Galvanized Steel
Microfiber, Stainless Steel
Polyester with Stainless Steel Channel
Stainless Steel
Width (English):
10 in.
14 in.
Description:
“Clean”
“Disinfect”
“Original”
“Universal”
Sterility:
Non-sterile
Sterile
12 product options available for selection
Product selection table with 12 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Material Width (English) Description Sterility
190321020 Stainless Steel 14 in. “Universal” Non-sterile
190321021 Galvanized Steel 14 in. “Universal” Non-sterile
190321022 Polyester with Stainless Steel Channel 14 in. “Disinfect” Non-sterile
190321023 Microfiber, Stainless Steel 14 in. “Clean” Non-sterile
190321005 Galvanized Steel 10 in. “Original” Non-sterile
190321000 Galvanized Steel 14 in. “Original” Non-sterile
189992406 Stainless Steel 14 in. “Original” Non-sterile
190321036 Stainless Steel 10 in. “Universal” Non-sterile
190321037 Galvanized Steel 10 in. “Universal” Non-sterile
190321030 Stainless Steel 10 in. “Original” Non-sterile
19200712 Stainless Steel 10 in. “Universal” Sterile
19200713 CE Universal Stainless Steel 14 in. “Universal” Sterile
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 12 options available.
12 options
Catalog No. 190321020 Supplier Vileda Professional™ Supplier No. 119654

Vileda Professional Roll-O-Matic™ sponge mop refills are made in the USA with Advanced Technology foam. They are manufactured specifically for the controlled environment industry, and allow 25% more liquid absorption and 2X more liquid release than competing foams.

Vileda Professional Roll-O-Matic™ sponge mop refills are made in the USA with Advanced Technology foam. They are manufactured specifically for the controlled environment industry, and allow 25% more liquid absorption and 2X more liquid release than competing foams.

All Vileda Professional brand refills are ideal for use with the "Pull and Lift" technique most commonly used in controlled environments. They are ideal for general use, absorbing spills, applying disinfectant, and cleaning particulates.

  • Fits all original and self-wringing Roll-O-Matic™ mops
  • 10- or 14-inch options for both stainless steel and galvanized steel handles
  • Completely autoclavable and resistant to gamma and ETO sterilization
  • Individually packaged and lot number controlled
  • Certificate of conformance and technical data available upon request
  • Sterile versions validated to 10-6 per ISO 11137, individually double bagged with lot number and indicator dot, and supplied with certificate of sterility upon request

Specifications

Product Type Roller Mop Refill
Color Blue
Sterility Non-sterile
Autoclavable Autoclavable
Width (English) 14 in.
Quantity 12/Cs.
For Use With (Equipment) Original and Self Wringing Roll-O-matic Roller Mops
Description “Universal”
For Use With (Application) For general use within all controlled environments, absorb spills, clean particulates, or apply disinfectants
Material Stainless Steel