Learn More
Vileda Professional™ Roll-O-Matic™ Self-Wringing Mops
Suitable for all controlled environments.
Supplier:  Vileda Professional™ 122910
 View more versions of this product
Catalog No. 190321024
Only null left
Description
Description
  • Mop with built-in wringer
  • Wringer built into handle, eliminating need for expensive and bulky wringer
  • Improved components eliminate sharp edges and provide for excellent refill retention
  • Fits all original and advanced technology Roll-O-Matic approved refills
  • Ergonomic grip
Specifications
Chemical Identifiers
Blue
For use on floors, walls, and ceilings in cleanroom environments. Ideal for the pull-and-lift cleaning technique
Specifications
58.5 to 76.25 x 14 in.
Sponge Mop Head, Straight
58.5 to 76.25 in.
14 in.
Roll-O-matic Self Wringing Cleanroom Mops
Aluminum
Extendable Mop
Blue
Not Autoclavable
Color Coding Tabs and Ergonomic Grip, Extendable Handle
For use on floors, walls, and ceilings in cleanroom environments. Ideal for the pull-and-lift cleaning technique
Product Suggestions
Product Suggestions