Learn More
Vileda Professional™ Cleanroom Squeegee with Swivel
Catalog No. 190321041
Change view
Click to view available options
Length (English):
20 in.
Catalog No. Length (English)
190321041 20 in.
1 options
Catalog No. 190321041 Supplier Vileda Professional™ Supplier No. 119933
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Swivels 360 degrees allowing for full range of motion
  • Use on floors and walls after applying disinfectant
  • Squeegee spills to the drain in controlled environments
  • Can be autoclaved and exposed to gamma or EtO sterilization
  • Use with FHP Extendable Handle #119966
Specifications
Product Type Swivel Squeegee
For Use With (Application) Cleanroom applications
Length (English) 20 in.