missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Vileda Professional™ Sieve for Pre-Prepared Mop Box
Description
- Fits on top of the Pre-Prepared Mop Box
- Evenly placed holes which ensure even saturation of mops
- Autoclavable
Specifications
Specifications
| Product Type | Pre-Mop Box Sieve |
| Color | Gray |
| Dimensions | 41 x 31 x 11 cm (16 x 12 x 5 in.) |
| For Use With (Equipment) | Pre-Mop Box |