Learn More
Vileda Professional™ CE Sweeper and Pan™
Catalog No. 19032171
Change view
Click to view available options
Product Type:
Dustpan Squeegee Sweeper
Open Lobby Dustpan
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type
19032171 Dustpan Squeegee Sweeper
19032172 Open Lobby Dustpan
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 19032171 Supplier Vileda Professional™ Supplier No. 119908
The CE Sweeper and Pan™ is ideal for ergonomic and efficient liquid removal. The system is hygienic, lightweight, and flexible. Perfect for dry or damp conditions.
  • Sweep up broken glass in your cleanroom quickly and easily
  • No need to change gloves when an item is dropped on the floor
  • Keeps workers from touching the floor within the controlled environment
  • Can be sprayed or wiped down with common disinfectants
  • Stands up easily for storage or can be hung
  • Adjustable bag holder for different size collection bags
Specifications
Product Type Dustpan Squeegee Sweeper
For Use With (Application) To sweep broken Glass, for the lobby dustpans and smooth floors where dirt could be dry or damp
For Use With (Equipment) Janitorial