missing translation for 'onlineSavingsMsg'
Learn More

Vileda Professional™ Buckets and Downpress Wringer Combination

Catalog No. 19200631
Change view
Click to view available options
Capacity (English):
6.5 to 10 gal.
6.5 to 8 gal.
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (English)
19200631 6.5 to 10 gal.
19200630 6.5 to 8 gal.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 19200631 Supplier Vileda Professional™ Supplier No. 131580

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Vileda Professional™'s bucket and wringer combos are designed to meet the demanding requirements of the most sanitary environments.

  • Quiet, smooth operation, easy to work with
  • More efficient wringing, avoids water spalshing on the floor
  • Accommodates all wet mop sizes with convenient opening
  • Durable, stain resistant one piece construction is easy to clean and sanitize and does not provide a breeding ground for bacteria
  • Downward pressure wringer forces water into bucket more effectively

Specifications

Product Type Bucket and Wringer
Capacity (Metric) 24.6 to 37.9 L
Material Plastic
Capacity (English) 6.5 to 10 gal.
Color Yellow
For Use With (Application) Designed to meet the demanding requirements of the most sanitary environments
For Use With (Equipment) Fits all mop sizes
Includes Gears and pressure plates with Fiber reinforced nylon, single bail carry handle, durable ball bearing casters, downward pressure wringer