Learn More
Vileda Professional™ 25L Longish Bucket With Color Coding Clips
For use on Controlled Environment Bucket Trolley or Pre-Prepared Trolley
Supplier: Vileda Professional™ 131662
Description
- For use on the CE Bucket Trolley or the CE Pre-Prepared Trolley
- Liquid measurement lines moldest inside the bucket
- Color coding clips included to allow differentiation in colors of bucket (blue, red, yellow,green)
- Polypropylene bucket with stainless steel handle
- Autoclavable
Specifications
| Longish Bucket | |
| Gray |