missing translation for 'onlineSavingsMsg'
Learn More

Labconco™ FreeZone™ Stoppering Tray Dryers, 230V Models

Catalog No. 10400419
Change view
Click to view available options
Plug Type:
North America
Includes:
3 Shelves
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Plug Type Includes
10400419 North America 3 Shelves
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 10400419 Supplier Labconco™ Supplier No. 794801010

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Includes

3 shelves

Requires

Freeze dryer, vacuum pump

Provides precise control and program flexibility for consistent performance. Labconco™ FreeZone™ Stoppering Tray Dryers, 230V Models are designed for lyophilizing moderate to large loads and stoppering vials or serum bottles under vacuum. They offer options for built-in 6-port manifold and isolation valve.

  • Three 16.5 x 12 in. temperature-controlled (±1° C), adjustable shelves with sample temperature sensors
  • Chamber pre-freezes samples to save time and money by eliminating need for separate freezer and product transfer
  • Up to 594 sq. inches of shelf space
  • Circulating fluid in shelves ranges from −40 and +40° C
  • Environmentally-friendly HCFC/CFC-free refrigeration system
  • Built-in pneumatic mechanism stoppers containers on all three shelves
  • Clear acrylic viewing door provides complete visibility of shelves
  • 1/8 in. OD backfill port introduces sterile air or inert gas from outside source to chamber, protecting samples from atmospheric moisture and contaminants
  • Lyo-Works™ OS provides real time display of freeze dryer and tray dryer parameters on 5 in. full color, capacitive touch screen on connected FreeZone 6, 12, or 18L Freeze Dryer (sold separately)
  • Data may be exported via USB or Ethernet
  • Microprocessor-based controller allows up to 30 different temperature programs to be stored and repeated, each using up to 16 different steps
  • Display provides navigation to manual or program mode, probe temperatures, vacuum in mBar, Torr or Pa, step of program in progress, time remaining in present step, and end of program, as well as alarms
  • Alerts and current operating parameter reading can be sent automatically to user-entered email addresses
  • Optional 6-Port Tray Dryer Manifold accommodates either 1/2 in. or 3/4 in. flask adapters and connect to flasks and other glassware
  • Optional built-in isolation valve separates Stoppering Tray Dryer from freeze dryer to shorten time necessary to reestablish working vacuum levels and to provide means for determining end point
  • Freeze dryer, vacuum pump with displacement of at least 163L/minute, and freeze dry glassware (if not bulk freeze drying) are required (not included)

Compliance

All models conform to UL Standard 61010-1 (60 Hz models), CAN/CSA C22.2 No. 61010-1 (60 Hz models), and CE Conformity marking (230V, 50 Hz models).

Warranty

One year

Specifications

Type Stoppering Tray Dryer
No. of Shelves 3
Temperature Range (Metric) -40°C to +40°C
Material Steel
Certifications/Compliance UL, CAN/CSA, CE
Includes 3 Shelves
Depth (English) 24.6 in.
Depth (Metric) 62.4 cm
Width (English) 32.4 in.
Width (Metric) 82.2 cm
Height (English) 27.1 in.
Height (Metric) 68.8 cm
Amperage 9A
Plug Type North America
Voltage 230 V
Frequency 60 Hz
Shipping Weight (English) 423 lb.
Shipping Weight (Metric) 191.86 kg
Show More Show Less