Learn More
Fisherbrand™ Freezer Storage Racks
Organize and optimize the freezers in the lab.
Supplier:  Fisherbrand™ 398279
 View more versions of this product
Catalog No. 03395489
Only null left
Description
Description
Organize and optimize the freezers in the lab
Make it easy to track samples by improving organization in your freezers. These racks enable you to store more samples, eliminate dead space, and minimize frost buildup.
  • Fit all standard types and sizes of upright and chest freezers, regardless of manufacturer
  • Store a variety of sample containers, including boxes, plates, and tubes
  • High-quality, corrosion resistant stainless steel
  • For use in all standard upright and chest freezers
Specifications
Chemical Identifiers
Not Autoclavable
Stainless Steel
Specifications
Not Autoclavable
Stainless Steel
Upright Freezers
55.88 cm
9.2 in.
13.97 cm
84
Plates
84 with lid, 96 without
84
22 in.
5.5 in.
Freezer Storage Rack
23.36 cm
Product Suggestions
Product Suggestions