Learn More
Thermo Scientific™ Freezer and Refrigerator Shelves
Catalog No. 361017651
Change view
Click to view available options
For Use With (Equipment):
23.3, 45.8, 51.1, 78.8 cu. ft. Units
29.2 cu. ft. Units
4.7 cu. ft., 11.5 cu. ft. Units
51.1 cu. ft. Units
Type:
Open Wire Shelves
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Equipment) Type
361017651 29.2 cu. ft. Units Open Wire Shelves
13990163 23.3, 45.8, 51.1, 78.8 cu. ft. Units Open Wire Shelves
361017654 51.1 cu. ft. Units Open Wire Shelves
12370653 4.7 cu. ft., 11.5 cu. ft. Units Open Wire Shelves
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 361017651 Supplier Thermo Scientific™ Supplier No. 6695
Maximize lab storage with Thermo Scientific™ Freezer and Refrigerator Shelves, designed for use as replacement shelves in high-performance laboratory refrigerators, freezers and ULTs.
Maximize lab storage with Thermo Scientific™ Freezer and Refrigerator Shelves, designed for use as replacement shelves in high-performance laboratory refrigerators, freezers and ULTs.
  • Epoxy-coated open-wire shelves
  • Full- and half-depth sizes
  • Undercounter units use full-depth shelves only
Specifications
Type Open Wire Shelves
Exterior Finish Epoxy-coated Steel
For Use With (Equipment) 29.2 cu. ft. Units
Warranty Warranty varies by country, please contact us for details
Holds General Freezer Items