Learn More
Research Products Upright Freezer Rack for 2 Inch High Boxes, 12 Box Capacity, 3 x 4 Array, 5 1/2 x 16 1/2 x 9 1/4H Inches DFS Item
Supplier:  Research Products Corporation RPI342
Convenient storage system for 1.5ml - 2.0ml micro-tubes and 1.2ml - 2.0ml cryogenic vials. Stainless steel freezer racks are available for this upright freeze storage system. Racks include stainless steel handles and accommodate 2 inch high cardboard and metal storage boxes.
Catalog No. 50212121
Only null left
Explore available promotions