missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ FreeStyle™ F17 Expression Medium

Catalog No. A1383504
Encompass
Change view
Click to view available options
Quantity:
1 L
10 L
20 L
6 x 1 L
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A1383504 20 L
50591354 1 L
50591355 6 x 1 L
A1383503 10 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. A1383504 Supplier Gibco™ Supplier No. A1383504

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

An animal origin-free, chemically defined, protein-free medium specifically developed to support the growth and transfection of 293-F and CHO cells in suspension without adaptation.

FreeStyle™ F17 Expression Medium is an animal origin–free, chemically defined, protein-free medium specifically developed to support the growth and transfection of 293-F and CHO cells in suspension without adaptation. This is a multi-platform medium compatible with a range of transient expression systems. The addition of 4–8 mM L-glutamine or GlutaMAX™ supplement is required before use. Gibco™ FreeStyle™ F17 Expression Medium features:

  • Little or no required adaptation
  • Ability to support suspension growth and transfection of 293-F and CHO cells
  • Animal origin–free, protein-free, and chemically defined formulation
  • Scalability in spinner flasks and bioreactors

    Little or no required adaptation
    Gibco™ 293-F, 293-H, FreeStyle™ 293-F, and CHO-S cells in suspension can be cultured directly into Gibco™ FreeStyle™ F17 Medium without adaptation. Cells adapted to other commercially available serum-free media can be subcultured directly into Gibco™ FreeStyle™ F17 Medium, usually without any further adaptation. Cells usually require adaptation from serum-containing formulations.

    Ability to support suspension growth and transfection of 293-F and CHO cells
    Gibco™ FreeStyle™ F17 Medium provides equal or better performance compared to Gibco™ FreeStyle™ 293 Expression Medium and Gibco™ FreeStyle™ CHO Medium. Gibco™ FreeStyle™ F17 Medium supports 293-F cells at a peak density of 6–7 × 106 viable cells per mL, and supports CHO-S cells at a peak density of 8–9 × 106 viable cells per mL. Cells cultured in Gibco™ FreeStyle™ F17 can be transfected using a wide variety of transfection reagents, including lipid-based transfection reagents, such as 293fectin™ and Freestyle™ MAX Reagent. Protein expression typically exceeds 75–150μg/mL IgG in 293-F cells.

    Animal origin–free, protein-free, and chemically defined formulation
    Gibco™ FreeStyle™ F17 Medium is animal origin–free, protein-free, and chemically defined, allowing for easier purification of your protein of interest. Gibco™ chemically defined media contain no proteins, hydrolysates, or components of unknown composition.

    Scalability in spinner flasks and bioreactors
    Protein production using Gibco™ FreeStyle™ F17 Medium can be scaled up in spinner flasks or bioreactors. The appropriate spinner or impeller speed and seeding density should be optimized for each system. Depending on the impeller design and speed, it may be necessary to supplement Gibco™ FreeStyle™ F17 Medium with additional Pluronic™ F-68 to avoid sheer stress in the culture.

    cGMP manufacturing and quality system
    Gibco™ FreeStyle™ F17 Medium is manufactured at a cGMP-compliant facility located in Grand Island, New York. The facility is registered with the FDA as a medical device manufacturer and is certified to ISO 13485 standards.
  • Specifications

    Cell Line HEK293
    Classification Animal Origin-free, Chemically-defined, Protein-free, Serum-free
    Concentration 1 X
    Form Liquid
    Product Type Expression Medium
    Serum Level Serum-free
    Sterility Sterile
    Without Additives No Glutamine
    For Use With (Application) Protein expression, viral vector production
    Product Line Freestyle
    Quantity 20 L
    Shipping Condition Room Temperature
    Endotoxin Level < 1 EU/mL
    Format Universal Bag(s)
    Purity or Quality Grade Qualified for Further Manufacturing
    Content And Storage Storage conditions: 2-8°C. Protect from light
    Shipping conditions: Ambient
    Shelf life: 12 months from date of manufacture
    Culture Environment CO2
    Culture Type Suspension Cell Culture
    Show More Show Less
    How long can I keep culturing FreeStyle cells after transfection?

    We typically keep the culture going for 7-10 days posttransfection. After that, protein expression may drop.

    What is the difference between FreeStyle 293 Expression Medium and FreeStyle F17 Expression Medium?

    FreeStyle F17 Expression Medium was designed from a different base formulation than FreeStyle 293 Expression Medium. FreeStyle F17 medium shows equal or better performance than the FreeStye 293 medium for certain cell lines and proteins. It is capable of supporting the growth of 293 and CHO cells and offers a wider variety and selection of transient cell culture media options.

    For Research use or Further Manufacturing. Not for diagnostic use or direct administration into humans or animals.