Learn More
Carver™ Lab Press Swivel Bearing Plates
Catalog No. 1387240
Change view
Click to view available options
Product Type:
Swivel Bearing Plate
Catalog No. Product Type
1387240 Swivel Bearing Plate
1 options
Catalog No. 1387240 Supplier Carver™ Supplier No. 2092#C
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Plate easily attaches to press for crushing and breaking tests on standard 2 in. (5cm) cubes or 2 × 4 in. (5 × 10cm) cylinders. Ball swivel exerts pressure evenly over entire sample surface.
Specifications
Product Type Swivel Bearing Plate
For Use With (Application) For crushing and breaking tests
For Use With (Equipment) For standard 2 in. cubes or 2 x 4 in. cylinders