Learn More
Carver™ Lab Press Cage Equipment
Catalog No. 1387210
Change view
Click to view available options
Product Type:
Cage Equipment
Catalog No. Product Type
1387210 Cage Equipment
1 options
Catalog No. 1387210 Supplier Carver™ Supplier No. 2094 #C
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
For pressing liquids out of semisolid materials. Complete with set of felt and steel discs, trays, bulb-type syringe and mounting hardware.
Specifications
Product Type Cage Equipment
For Use With (Application) To press moisture from fibrous materials, extract fats, oils or juices from vegetable or animal tissues, splitting chilled oils or separating crystals from liquors
For Use With (Equipment) For carver lab press
Includes Cylinder, plunger, steel discs, felt discs, tray and bulb syringe, mounting hardware