missing translation for 'onlineSavingsMsg'
Learn More

FRAP1 (mTOR) Assay Buffer

Catalog No. PV4794
Encompass
Change view
Click to view available options
Quantity:
20 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PV4794 20 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PV4794 Supplier Thermo Fisher Scientific Supplier No. PV4794

FRAP1 (mTOR) Assay Buffer

1X Assay Buffer for use with the LanthaScreen™ FRAP1 (mTOR) assays.

Specifications

Chemical Name or Material Buffer Solution
For Use With (Equipment) LanthaScreen™ Nuclear Receptor Cofactor Interaction Assays
Physical Form Liquid
Quantity 20 mL

For Research Use Only. Not for use in diagnostic procedures.