missing translation for 'onlineSavingsMsg'
Learn More

Bulwark™ FR Lab Coats, Knit Cuffs

Catalog No. 17004379
Change view
Click to view available options
Size:
X-Small
Small
Medium
Large
X-Large
2X-Large
3X-Large
4X-Large
5X-Large
Style:
Regular
Tall
16 product options available for selection
Product selection table with 16 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size Style
17004379 5X-Large Tall
17004364 X-Small Regular
17004365 Small Regular
17004366 Medium Regular
17004367 Large Regular
17004368 X-Large Regular
17004369 2X-Large Regular
17004370 3X-Large Regular
17004371 4X-Large Regular
17004372 5X-Large Regular
17004373 Medium Tall
17004374 Large Tall
17004375 X-Large Tall
17004376 2X-Large Tall
17004377 3X-Large Tall
17004378 4X-Large Tall
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 16 options available.
16 options
Catalog No. 17004379 Supplier Bulwark™ Supplier No. KNC2RB 5L 0L

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Lab coat designed for functionality, comfort, and safety with a CAT 1 arc rating and NFPA 2112 compliance.

  • Certifications Level 1: NFPA 2112, ASTM F1506
  • Certifications Level 2: Category 1 Protection
  • Hazard: Arc Flash, Flash Fire
  • Wash Care: Industrial Laundry - Light Soil
  • Rib cuffs for protection during movement
  • Concealed snap front closure to avoid scratching surfaces
  • Four front mitered style pockets for ample storage
  • Pass-through at hips to access underneath
  • Protection: ATPV Arc Rating: 4.4
  • Country of Origin: Imported, Made in USA Fabric

Specifications

Product Type Flame-resistant Lab Coat
Size 5X-Large
Gender Men's
Material Nomex
Color Royal Blue
Closure Type Snaps
Cuff Style Knit
Disposable Reusable
Sterility Non-sterile
Resistance Flame
Style Tall
No. of Pockets 4
Model KNC2RB
Arc Flash Rating 4.4 Calories/cm2
Certifications/Compliance NFPA 2112, ASTM 1506
Construction Sewn
Show More Show Less