missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bulwark™ FR Lab Coats, Knit Cuffs
Lab coat designed for functionality, comfort, and safety with a CAT 1 arc rating and NFPA 2112 compliance.
Supplier: Bulwark™ KNC2RB4L0R
Description
- Certifications Level 1: NFPA 2112, ASTM F1506
- Certifications Level 2: Category 1 Protection
- Hazard: Arc Flash, Flash Fire
- Wash Care: Industrial Laundry - Light Soil
- Rib cuffs for protection during movement
- Concealed snap front closure to avoid scratching surfaces
- Four front mitered style pockets for ample storage
- Pass-through at hips to access underneath
- Protection: ATPV Arc Rating: 4.4
- Country of Origin: Imported, Made in USA Fabric
Specifications
| Flame-resistant Lab Coat | |
| Men's | |
| Royal Blue | |
| Knit | |
| Non-sterile | |
| Regular | |
| KNC2RB | |
| NFPA 2112, ASTM 1506 |
| 4X-Large | |
| Nomex | |
| Snaps | |
| Reusable | |
| Flame | |
| 4 | |
| 4.4 Calories/cm2 | |
| Sewn |