Learn More
International Enviroguard™ FR and Chemical Resistant Coverall with Attached Hood and Elastic Wrists
PyroGuard CRFR flame-resistant coveralls are designed to wear over your primary FR garments, helping to extend the life of your primary FR clothing.
$383.49 - $499.90
Specifications
Product Type Flame-resistant Coveralls
Gender Unisex
Material CFR Bilaminate Fabric
Color Gray
Disposable Single-use
View More Specs
Products 8
Catalog Number Mfr. No. Size Price Quantity  
Catalog Number Mfr. No. Size Price Quantity  
17-184-014
View Documents
International Enviroguard™
9115TM
Medium
Case of 6 Each for $383.49
Only null left
17-184-015
View Documents
International Enviroguard™
9115TL
Large
Case of 6 Each for $385.30
Only null left
17-184-013
View Documents
International Enviroguard™
9115TXL
X-Large
Case of 6 Each for $391.44
Only null left
17-184-020
View Documents
International Enviroguard™
9115T2XL
2X-Large
Case of 6 Each for $402.74
Only null left
17-184-019
View Documents
International Enviroguard™
9115T3XL
3X-Large
Case of 6 Each for $426.44
Only null left
17-184-018
View Documents
International Enviroguard™
9115T4XL
4X-Large
Case of 6 Each for $451.24
Only null left
17-184-017
View Documents
International Enviroguard™
9115T5XL
5X-Large
Case of 6 Each for $480.30
Only null left
17-184-016
View Documents
International Enviroguard™
9115T6XL
6X-Large
Case of 6 Each for $499.90
Only null left
Description
Description
  • Coveralls also offer protection from a broad range of chemicals and provide flash fire protection that meets NFPA 2113
  • Advanced, self-extinguishing fabric does not melt or drip when ignited to prevent flames from spreading
  • Attached respirator-fit hood combined with secure, elastic wrists accommodate a respirator to prevent toxic inhalation and keep sleeves in place to avoid skin exposure
  • PyroGuard CRFR flame-resistant suits stop flames and chemicals in their tracks offering the comfort and safety you need to work in tough environments
  • Proprietary bilaminate fabric protects against flames and chemicals
  • Elastic back stretches with user for more comfort and better tear resistance
  • Storm flap over zipper adds an extra layer of protection
  • Taped seams offer added protection over serged seam coveralls
  • Possible applications include petrochemical environments, manufacturing, industrial, oil/gas, and more
Specifications
Specifications
Flame-resistant Coveralls
CFR Bilaminate Fabric
Single-use
Elastic
Attached, Elastic Face
Serged, Taped
Acid, Anti-static, Caustics, Chemicals, Flame
None
Unisex
Gray
Elastic at Back
Hooded
2-way Front Zipper with Storm Flap
Non-sterile
Yes
Videos
SDS
missing translation for 'documents'
missing translation for 'documents'