missing translation for 'onlineSavingsMsg'
Learn More

MP Biomedicals FP96 TeenFlex Metal Adapter

Catalog No. MP116010675
Change view
Click to view available options
Holds:
12 x 15 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Holds
MP116010675 12 x 15 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MP116010675 Supplier MP Biomedicals Supplier No. 116010675
Only null left

Securely holds up to 12 x 15 mL tubes. Resistant to a wide range of temperatures, suitable for various sample preparation conditions.

  • The medium-scale FP96 TeenFlex Metal Adapter for the FastPrep-96™ holds 12 x 15 mL Lysing Matrix tubes and is compatible with the SafTest™ Food Inspection System.
  • It allows frozen samples stored in Lysing Matrix tubes at -20°C or -80°C to be immediately processed with minimal hands-on manipulation, helping to prevent degradation of cellular components by endogenous enzymes.
  • A wide range of disposable 15 mL Lysing Matrix tubes ensures thorough homogenization of virtually any sample type in seconds.

Specifications

Product Type Adapter
Blade Material Stainless Steel
Color Blue
For Use With (Equipment) FastPrep-96, FastPrep-96 Pro
Holds 12 x 15 mL