missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MP Biomedicals FP96 LargeFlex Metal Adapter
Description
- Super-sized FP96 LargeFlex Metal Adapter for the FastPrep-96™ and FastPrep-96™ Pro that holds 2 x 250 mL Lysing Matrix Bottles.
- For scale-up and small production run capabilities
- Compatible with the SafTest™ Food Inspection Platform.
- The ergonomic design of the adapter ensures easy loading of Lysing Matrix bottles that remain securely sealed during the homogenization.
- Additionally, frozen samples in bottles loaded in the adapter and stored at -20°C or -80°C are ready to be immediately processed with minimal hands-on manipulation, preventing degradation of cellular components by endogenous enzymes.
- A wide range of disposable 250 mL Lysing Matrix bottles ensure thorough homogenization of any sample type in seconds.
- Made from durable anodized aluminum for long-lasting performance
Specifications
Specifications
| Product Type | Adapter |
| Blade Material | Stainless Steel |
| Color | Blue |
| For Use With (Equipment) | FastPrep-96, FastPrep-96 Pro |
| Holds | 2 x 250 mL |