Learn More
S-Curve™ Four Compartment Glove Dispenser with Single Lid
Catalog No. 19160199
Change view
Click to view available options
Material:
Acrylic
PETG
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Material
19160199 Acrylic
19162429 PETG
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 19160199 Supplier S-Curve™ Supplier No. GD405
Only null left
Four compartment dispenser includes a single, sloping lid with a full-length hinge to maintain HEPA flow.
  • Single, sloping lid with full-length hinge to maintain HEPA flow
  • Can be mounted with heavy-duty wall brackets (included)
  • Flame treated edges for a smooth finish
Specifications
Product Type Glove Dispenser
Color Clear
Dimensions (D x W x H) Interior 12 x 21 x 12 in.
Holds Gloves
Material Acrylic
Mounting Keyhole Slots
No. of Compartments 4
Product Style Top Loading
Includes Heavy-duty wall mounting bracket
Thickness (English) 0.25 in.