missing translation for 'onlineSavingsMsg'
Learn More

Forskolin, 98+%

Catalog No. aaj63292ma
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ63292MA 10 mg
AAJ63292MF 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAJ63292MA Supplier Thermo Scientific Chemicals Supplier No. J63292MA
Only null left

Forskolin, CAS # 66575-29-9, is a diterpenoid compound isolated from Indian Coleus that acts as a strong activator of the adenylate cyclase system. | CAS: 66575-29-9 | C22H34O7 | 410.507 g/mol

Forskolin acts as an adenylyl cyclase activator. It is used in the treatment of hypertension, asthma, eczema, psoriasis, congestive heart failure, angina, stomachache, gastric and intestinal spasm. It also plays an important role in the treatment of rheumatism, heart, blood and circulation conditions and cancers. It is used to induce menstruation and as an oral contraceptive.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General Description

• Forskolin is a diterpenoid and cyclic ketone, found naturally as a plant metabolite

• It is a protein kinase A agonist and a cyclic ketone • Forskolin is an antihypertensive and muscle relaxant; it can also promote the release of pituitary hormones and prevent platelet aggregation

Application

• Forskolin assists in neuron differentiation in vitro

• Useful for studies of cell metabolism and development; there is some evidence of its usefulness in the study of glaucoma

• It is intended for cell culture and biochemistry applications

Chemical Identifiers

CAS 66575-29-9
Molecular Formula C22H34O7
Molecular Weight (g/mol) 410.507
MDL Number MFCD00082317
InChI Key OHCQJHSOBUTRHG-KGGHGJDLSA-N
Synonym forskolin, colforsin, coleonol, colforsina, colforsine, colforsinum, boforsin, colforsine french, colforsinum latin, colforsina spanish
PubChem CID 47936
ChEBI CHEBI:42471
IUPAC Name [(3R,4aR,5S,6S,6aS,10S,10aR,10bS)-3-ethenyl-6,10,10b-trihydroxy-3,4a,7,7,10a-pentamethyl-1-oxo-5,6,6a,8,9,10-hexahydro-2H-benzo[f]chromen-5-yl] acetate
SMILES CC(=O)OC1C(C2C(CCC(C2(C3(C1(OC(CC3=O)(C)C=C)C)O)C)O)(C)C)O

Specifications

Color White
Quantity 10 mg
Beilstein 1692716
Merck Index 14,2476
Solubility Information Soluble in ethanol,dimethylsulfoxide and chloroform; Insolule in water.
Formula Weight 410.51
Percent Purity ≥98%
Physical Form Solid
Chemical Name or Material Forskolin
Hazard Category H312
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P280-P302+P352-P312-P363-P501c
EINECSNumber 266-410-9
RTECSNumber QL6150000
TSCA No
Recommended Storage Store at -20°C

RUO – Research Use Only