missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Formoxime (10% in Water, ca. 2.4mol/L), TCI America™
Specifications
Specifications
| Color | Colorless |
| Quantity | 25 mL |
| Formula Weight | 45.04 |
| Physical Form | Liquid |
| Chemical Name or Material | Formoxime (10% in Water, ca. 2.4mol/L) |
Safety and Handling
| RTECSNumber | LP9720000 |
| TSCA | Yes |
| Recommended Storage | Refrigerator |