missing translation for 'onlineSavingsMsg'
Learn More

Formoterol Fumarate Hydrate, TCI America™

Catalog No. F088110MG
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F088110MG 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. F088110MG Supplier TCI America Supplier No. F088110MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 43229-80-7
Molecular Formula C42H52N4O12
Molecular Weight (g/mol) 804.894
MDL Number MFCD03093863
InChI Key OBRNDARFFFHCGE-QDSVTUBZSA-N
Synonym formoterolfumarate, aformoterol, atock, foradil aerolizer, foradil certihaler, formoterol fumarate, formoterol hemifumarate, foradil acrolizer, oxis pmdi
PubChem CID 53477580
ChEBI CHEBI:63108
IUPAC Name (E)-but-2-enedioic acid;N-[2-hydroxy-5-[(1R)-1-hydroxy-2-[[(2R)-1-(4-methoxyphenyl)propan-2-yl]amino]ethyl]phenyl]formamide
SMILES CC(CC1=CC=C(C=C1)OC)NCC(C2=CC(=C(C=C2)O)NC=O)O.CC(CC1=CC=C(C=C1)OC)NCC(C2=CC(=C(C=C2)O)NC=O)O.C(=CC(=O)O)C(=O)O

Specifications

Color White-Yellow
Quantity 10 mg
Formula Weight 402.45
Physical Form Crystalline Powder
Chemical Name or Material Formoterol Fumarate Hydrate
RTECSNumber LQ2988000
TSCA No
Recommended Storage Freezer