Learn More
StatLab™ Formical-2000 Decalcifier
Catalog No. STL131432
Change view
Click to view available options
Quantity:
1 Gallon
5 gal. Cube
1 Quart
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
STL131432 1 Quart
STL13141 1 Gallon
STL13145 5 gal. Cube
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Harmful if swallowed, in contact with skin, or inhaled.
Catalog No. STL131432 Supplier StatLab™ Supplier No. 131432
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Harmful if swallowed, in contact with skin, or inhaled.
A fast yet gentle bone decalcifier.
  • Formulated from semiconductor grade formic acid, highly purified reagent grade water and EDTA
  • Works for all routine histological specimens, as well as IHC
  • Will safely decalcify bone without destroying antigen sites
Specifications
Quantity 1 Quart
Solution Type Decalcifier