missing translation for 'onlineSavingsMsg'
Learn More

Formic Acid Solution, Honeywell Fluka™

Catalog No. 6000605
Change view
Click to view available options
Quantity:
100 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
6000605 100 mL
6000606 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 6000605 Supplier Honeywell Chemicals Supplier No. 09676100ML
Only null left

Puriss. p.a., for HPLC, 50% in water, 49-51% (T)

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured

Chemical Identifiers

CAS 64-18-6
Molecular Formula CH2O2
Molecular Weight (g/mol) 46.025
MDL Number MFCD00003297
InChI Key BDAGIHXWWSANSR-UHFFFAOYSA-N
Synonym methanoic acid, formylic acid, aminic acid, bilorin, hydrogen carboxylic acid, formisoton, myrmicyl, formira, acide formique, collo-bueglatt
PubChem CID 284
ChEBI CHEBI:30751
IUPAC Name formic acid
SMILES C(=O)O

Specifications

Linear Formula HCOOH
UN Number 3412
Beilstein 1209246
Concentration 49 to 51% (T)
Formula Weight 46.03g/mol
Grade puriss. p.a.
Quantity 100 mL
Chemical Name or Material Formic Acid Solution
Hazard Statement P261-P280-P301 + P312 + P330-P303 + P361 + P353-P304 + P340 + P310-P305 + P351 + P338
Precautionary Statement H302 + H332-H314